missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) oxalate trihydrate, 98%

Product Code. 11449377
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449377 100 g 100g
11459377 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449377 Supplier Thermo Scientific Alfa Aesar Supplier No. B21411.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13268-42-3 | C6H18FeN3O15 | 428.06 g/mol

Ammonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Light sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13268-42-3
Molecular Formula C6H18FeN3O15
Molecular Weight (g/mol) 428.06
MDL Number MFCD00150137
InChI Key ARJSTCBPHIPXIJ-UHFFFAOYSA-K
Synonym Ferric ammonium oxalate; Iron(III) ammonium oxalate
IUPAC Name triammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4⁵.4⁵]tridecane-5,5,5-tris(ylium) trihydrate
SMILES [NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4

Specifications

Quantity 100 g
UN Number UN3288
Merck Index 14,517
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 428.07 (374.02 Anhydrous)
Percent Purity 98%
Sensitivity Hygroscopic; Light sensitive
Density 1.78
Chemical Name or Material Ammonium iron(III) oxalate trihydrate
compliance-icons
Product Identifier
  • Ammonium iron(III) oxalate trihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.