missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) sulfate dodecahydrate, 98+%

Product Code. 11443137
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
This item is not returnable. View return policy

Product Code. 11443137

Brand: Thermo Scientific Alfa Aesar A16105.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-83-7 | FeH28NO20S2 | 482.176 g/mol

Ammonium iron(III) sulfate dodecahydrate is a reagent that is used as a mordant in dyeing and textile printing. It is also utilized in analytical chemistry as an iron standard. In biochemistry, ammonium ferric sulfate is used as a catalyst for the generation of free radicals. Increased xanthine oxidase and xanthine oxidoreductase activity in cultured rat cells has been demonstrated in the presence of ammonium ferric sulfate. It may be used as an iron source to restore enzymatic activity in apoenzymes, such as for soybean lipoxygenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iron(III) sulfate dodecahydrate is a reagent that is used as a mordant in dyeing and textile printing. It is also utilized in analytical chemistry as an iron standard. In biochemistry, ammonium ferric sulfate is used as a catalyst for the generation of free radicals. Increased xanthine oxidase and xanthine oxidoreductase activity in cultured rat cells has been demonstrated in the presence of ammonium ferric sulfate. It may be used as an iron source to restore enzymatic activity in apoenzymes, such as for soybean lipoxygenase.

Solubility
Soluble in water (1240 mg/ml at 25°C) and 1% (v/v) aqueous HCl (100 mg/ml). Insoluble in alcohol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
MDL Number MFCD00150004
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Melting Point 39°C to 41°C
Quantity 500 g
Linear Formula NH4Fe(SO4)2·12H2O
Merck Index 14,521
Solubility Information Soluble in water (1240mg/ml at 25°C) and 1% (v/v) aqueous HCl (100mg/ml). Insoluble in alcohol.
Formula Weight 482.18 (266.00 Anhydrous)
Percent Purity ≥98%
Odor Odorless
Density 1.71
Chemical Name or Material Ammonium iron(III) sulfate dodecahydrate
compliance-icons
Product Identifier
  • Ammonium iron(III) sulfate dodecahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Iron(III) ammonium sulfate dodecahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.