missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) sulfate dodecahydrate, ACS, 98.5-102.0%

Product Code. 11328178
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328178 500 g 500g
11318178 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328178 Supplier Thermo Scientific Alfa Aesar Supplier No. 039391.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7783-83-7 | FeH28NO20S2 | 482.176 g/mol

Ammonium iron(III) sulfate dodecahydrate is used in dyeing and printing textiles, as analytical lab reagent. It is often employed as an analytical standard, and has been used in a variety of other applications from nanomaterials to general redox reactions. It plays an important role for the inactivation of mycobacteriophage D29 bacteria which is vivale for viable mycobacterium smegmatis and m.tuberculosis. The crystalline ammonium ferrous sulfate has shown useful function as the high absorbed dose and low-temperature dosimeter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
MDL Number MFCD00150004
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Melting Point 39°C to 41°C
Color Purple
Physical Form Broken Crystals
Quantity 500 g
Odor Odorless
Appearance Pale purple broken crystals
Assay Percent Range 98.5-102.0%
Density 1.71 g/cm3
Solubility Information Freely soluble in water and hydrochloric acid. Insoluble in alcohol.
Formula Weight 482.18 (266.00 Anhydrous)
Grade ACS
Chemical Name or Material Ammonium iron(III) sulfate dodecahydrate, For analysis ACS
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium iron(III) sulfate dodecahydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P362-Take off contaminated clothing.
  • P321-Specific treatment (see on this label).
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.