missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) sulfate dodecahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-102.0%, crystalline, slightly violet, Solstice

Product Code. 15623230
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Unit Size:
2.5kg
500g
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15623230 500 g 500g
15613230 2.5 kg 2.5kg
15633230 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15623230 Supplier Honeywell-Fluka Supplier No. 31111500G
  • ONLINE EXCLUSIVE PRICE
    55.04 €
    51.52 € / 500g
    Save 3.52 €
    6% Off
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-102.0%, crystalline, slightly violet

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Color Violet
Physical Form Crystalline
Quantity 500 g
Linear Formula NH4Fe(SO4)2 · 12H2O
MDL Number MFCD00150004
UN Number NONH for all modes of transport
Solubility Information In accordance with water, 10g/dL
Formula Weight 482.19g/mol
Percent Purity 99 to 102%
Grade ACS/puriss. p.a.
Chemical Name or Material Ammonium Iron(Iii) Sulfate Dodecahydrate, Reag. Ph. Eur. reag. ISO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.