missing translation for 'onlineSavingsMsg'
Learn More

Ammonium metavanadate, Solstice

Product Code. 16685793
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16685793 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16685793 Supplier Fluka Supplier No. 3115325KG
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

Melting Point 200°C (Decomposes before melting)
CAS 7803-55-6
Color Light Yellow
CAS Max % 1
pH 7
Quantity 25 kg
Molecular Formula H4NO3V
Linear Formula NH4VO3
MDL Number MFCD00011430
Synonym Ammonium vanadate(V)
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
SMILES N.O[V](=O)=O
IUPAC Name vanadiumoylol amine
Molecular Weight (g/mol) 116.98
Grade ACS
Density 2.3 g/cm3 (20°C)
Chemical Name or Material Ammonium metavanadate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.