missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate, 99.99% (metals basis)

Product Code. 11399268
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11399268

Brand: Thermo Scientific Alfa Aesar 043206.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13106-76-8 | H8MoN2O4

It is used to prepare phosphomolybdates, to measure the amount of phophates, silicates, arsenates, lead in pigments, river water and sea water. It is also used as a negative stain in electron microscopy, in photogrphy, ceramics, paints. It is used in the production of molybdenum. It is used in the preparation of dehydrogenation and desulphurization catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13106-76-8
Molecular Formula H8MoN2O4
MDL Number MFCD00011596

Specifications

Color White
Physical Form Lump/Powder
Quantity 100 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in water,acid and alkali,but insoluble in alcohol.
Formula Weight 196.02
Odor Odorless
Chemical Name or Material Ammonium molybdate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.