missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate (di), Mo 56.5%

Product Code. 11336628
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11336628 250 g 250g
11316628 50 g 50g
11326628 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11336628 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. 012215.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 27546-07-2 | H2MoO4 | 161.96 g/mol

Ammonium molybdate (di) is used as pigments, agricultural fertilizer, color lakes and fabric fire retardant agent. It is also used as a raw material for the production of molybdenum wire, ceramic glazes and other molybdenum compounds. Further, it is used in photography, ceramics and paints. It is also used reagent for analysis as well as in the determination of phosphorus in analytical chemistry. In addition, it is used as a catalyst in the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 27546-07-2
Molecular Formula H2MoO4
Molecular Weight (g/mol) 161.96
MDL Number MFCD00010882
InChI Key VLAPMBHFAWRUQP-UHFFFAOYSA-L
Synonym Ammonium dimolybdate

Spezifikation

Physical Form Powder
Quantity 250 g
Solubility Information Soluble in water.
Formula Weight 339.95
Concentration or Composition (by Analyte or Components) Mo 56.5%
Chemical Name or Material Ammonium molybdate (di)

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.