missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Molybdate Tetrahydrate, puriss. p.a. ACS Reagent, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 500 g |
| Linear Formula | (NH4)6Mo7O24 · 4H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 1235.86g/mol |
| Percent Purity | ≥99% (T) |
| Grade | ACS/puriss. p.a. |
| Packaging | Plastic Bottle |
| Density | 2.498g/mL at 25°C |
| Chemical Name or Material | Ammonium Molybdate Tetrahydrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?