missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium molybdate(VI) tetrahydrate, 99+%, for analysis
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysis
Specifications
Specifications
| Melting Point | 190.0°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Assay Percent Range | 99% min |
| Linear Formula | (NH4)6Mo7O24·4H2O |
| Merck Index | 15,53 |
| Solubility Information | Solubility in water: 400g/L (20°C). Other solubilities: practically insoluble in alcohol |
| Formula Weight | 1235.86 |
| Percent Purity | ≥99% |
| Show More |
Safety and Handling
Product Identifier
- Ammonium molybdate(VI) tetrahydrate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Ammonium molybdate(VI) tetrahydrate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur