missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate(VI) tetrahydrate, ACS reagent

Product Code. 10792592 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10792592 500 g Glass bottle 500g
10792402 5 g Glass bottle 5g
10773223 100 g Plastic bottle 100g
10562714 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10792592 Supplier Thermo Scientific Acros Supplier No. 423315000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12054-85-2 | H24Mo7N6O24·4H2O | 1235.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-85-2
Molecular Formula H24Mo7N6O24·4H2O
MDL Number MFCD00167059

Specifications

Color White
Physical Form Crystalline Powder
Quantity 500 g
Linear Formula (NH4)6Mo7O24·4H2O
Merck Index 15, 530
Formula Weight 1235.86
Percent Purity 81.0 to 83.0% (MoO3)
Grade ACS Reagent
Packaging Glass bottle
Decomposition Information >150°C
Density 2.4900g/mL
Chemical Name or Material Ammonium molybdate(VI) tetrahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium molybdate(VI) tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium molybdate(VI) tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.