missing translation for 'onlineSavingsMsg'
Learn More

Ammonium O,O'-dimethyldithiophosphate, 95%

Product Code. 11374666
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374666 1 g 1g
11364666 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374666 Supplier Thermo Scientific Alfa Aesar Supplier No. H32592.03
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1066-97-3 | C2H10NO2PS2 | 175.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-97-3
Molecular Formula C2H10NO2PS2
Molecular Weight (g/mol) 175.201
MDL Number MFCD09753116
InChI Key PPGORMGERPBFTJ-UHFFFAOYSA-N
Synonym ammonium o,o-dimethyl dithiophosphate, ammonium o,o'-dimethyldithiophosphate, ammonium o,o-dimethyl sulfanidylphosphonothioate, ammonium dimethyl phosphorodithioate, ammonium o,o'-dimethylphosphorodithioate, azanium dimethoxy-sulfanylidene-sulfido, o,o-dimethylphosphorodithioic acid ammonium salt, ammonium dimethoxy-sulfido-thioxo-$l^ 5-phosphane, azanium;dimethoxy-sulfanylidene-sulfido-?^ 5-phosphane
PubChem CID 6451175
IUPAC Name azanium;dimethoxy-sulfanylidene-sulfido-$l^{5}-phosphane
SMILES COP(=S)(OC)[S-].[NH4+]

Specifications

Melting Point 146°C to 150°C
Odor Weak
Quantity 1 g
Formula Weight 175.21
Percent Purity 95%
Chemical Name or Material Ammonium O,O'-dimethyldithiophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.