missing translation for 'onlineSavingsMsg'
Learn More

Ammonium oxalate monohydrate, 99+%, ACS reagent

Product Code. 10054013 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10054013 100 g 100g
10697211 500 g 500g
10552754 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10054013 Supplier Thermo Scientific Acros Supplier No. 423361000
  • ONLINE EXCLUSIVE PRICE
    28.35 €
    27.50 € / 100g
    Save 0.85 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6009-70-7 | C2H8N2O4·H2O | 142.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H8N2O4·H2O
Molecular Weight (g/mol) 142.11
MDL Number MFCD00149694
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

Melting Point 70.0°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99+%
Linear Formula (CO2NH4)2·H2O
Merck Index 15, 534
Solubility Information Solubility in water: 45g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 142.11
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic bottle
Residue after Ignition 0.02% max
Chemical Name or Material Ammonium oxalate monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium oxalate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.