missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Oxalate Monohydrate, Extra Pure, SLR, Fisher Chemical

Product Code. 10283270 Shop All Fisher Chemical Products
Change view
Click to view available options
500g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10283270 500g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10283270 Supplier Fisher Chemical Supplier No. A/6000/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 70°C
CAS 6009-70-7
Color White
pH 6.4
Physical Form Solid
Quantity 500 g
Molecular Formula C2H10N2O5
MDL Number 149694
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
SMILES N.N.O.OC(=O)C(O)=O
IUPAC Name oxalic acid diamine hydrate
Molecular Weight (g/mol) 142.11
Formula Weight 142.11g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium oxalate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium oxalate, monohydrate, Ammoniumoxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.