missing translation for 'onlineSavingsMsg'
Learn More

Ammonium peroxydisulfate, ACS, 98.0% min

Product Code. 11478660
Change view
Click to view available options
Quantity:
100 g
1 kg
5 x 1 kg
Unit Size:
100g
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478660 5 x 1 kg 5kg
11468660 100 g 100g
11458660 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478660 Supplier Thermo Scientific Alfa Aesar Supplier No. 054106.D9
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol

Ammonium peroxydisulfate is used as an oxidizer, a bleaching agent, in the field of photography, to make aniline dyes, electroplating and in analytical chemistry for determination of manganese.Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diazanium;sulfonatooxy sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C (decomposition)
Physical Form Lump
Quantity 5 x 1 kg
Flash Point None
Refractive Index 1.5
Sensitivity Moisture sensitive
Assay Percent Range 98.0% min
Density 1.982
Solubility Information Freely soluble in water.
Formula Weight 228.19
Grade ACS
Chemical Name or Material Ammonium peroxydisulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.