missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, for Electrophoresis, Fisher Chemical

Product Code. 10744171 Shop All Fisher Chemical Products
Change view
Click to view available options
:
25g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10744171 25g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10744171 Supplier Fisher Chemical Supplier No. A/P470/46
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 120°C
CAS 7727-54-0
Color White
pH 3.2
Physical Form Powder/Solid
Quantity 25 g
Molecular Formula H8N2O8S2
MDL Number 3390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular Weight (g/mol) 228.19
Formula Weight 228.19g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.