missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10223230 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 kg
3 kg
Unit Size:
1kg
3kg
This item is not returnable. View return policy

Product Code. 10223230

Brand: Fisher Chemical A/6120/60

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number 3390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C
Color White
pH 3.2
Physical Form Powder/Solid
Quantity 1 kg
Formula Weight 228.19g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.