missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, Solstice

Product Code. 15603240 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
6 x 1 kg
Unit Size:
1kg
5kg
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15603240 5 kg 5kg
15693230 1 kg 1kg
15613240 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15603240 Supplier Honeywell-Fluka Supplier No. 311175KG
  • ONLINE EXCLUSIVE PRICE
    291.00 €
    262.25 € / 5kg
    Save 28.75 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Quantity 5 kg
Linear Formula (NH4)2S2O8
UN Number 1444
Solubility Information Soluble in water
IUPAC Name diazanium;sulfonatooxy sulfate
Formula Weight 228.2g/mol
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Residue after Ignition ≤0.05% (as SO4)
Density Vapor Density: 7.9 (vs air)
Chemical Name or Material Ammonium Persulfate, Reag. Ph. Eur.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium persulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: diammonium peroxodisulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.