missing translation for 'onlineSavingsMsg'
Learn More

Ammonium phosphate, dibasic, 98+%, ACS reagent

Product Code. 10445765 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10445765 2.5 kg Plastic bottle 2.5kg
10197650 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10445765 Supplier Thermo Scientific Acros Supplier No. 423370025
  • ONLINE EXCLUSIVE PRICE
    190.00 €
    184.00 € / 2.5kg
    Save 6.00 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
SMILES N.N.OP(O)(O)=O

Specifications

Color White
pH 7.7 to 8.1 (5% soln. at 25°C)
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 2.5 kg
Assay Percent Range 98+%
Linear Formula (NH4)2HPO4
Merck Index 15, 540
Solubility Information Solubility in water: 58g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone
IUPAC Name phosphoric acid diamine
Formula Weight 132.06
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Density 1.61
Chemical Name or Material Ammonium phosphate, dibasic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.