missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Dibasic, TraceSELECT™, ≥99.995% (Metals basis), Honeywell™ Fluka™

Product Code. 15645520 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Poly bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15645520 25 g Poly bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15645520 Supplier Honeywell-Fluka Supplier No. 4339125G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 155°C
CAS 7783-28-0
Color White
pH 7.5 to 9.0 (at 132.10g/L 25°C)
Physical Form Crystalline
Quantity 25 g
Molecular Formula H9N2O4P
Linear Formula (NH4)2HPO4
MDL Number MFCD00010891
UN Number NONH for all modes of transport
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILES N.N.OP(O)(O)=O
IUPAC Name diazanium;hydrogen phosphate
Molecular Weight (g/mol) 132.06
PubChem CID 24540
ChEBI CHEBI:63051
Formula Weight 132.06g/mol
Percent Purity ≥99.995% (metals basis)
Grade TraceSELECT
Odor Odorless
Packaging Poly bottle
Density ∽1.62g/cu.cm at 20°C
Chemical Name or Material Ammonium phosphate dibasic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.