missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Purpurate, Pure, Metal Indicator, Fisher Chemical™

Product Code. 10293270 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10293270 5 g 5g
10519760 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10293270 Supplier Fisher Chemical Supplier No. A/6200/44

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 3051-09-0
Molecular Formula C8H8N6O6
Molecular Weight (g/mol) 284.19
MDL Number 12777
InChI Key PCCZWUCAYILEMX-UHFFFAOYSA-O
IUPAC Name ammonium 2,4,6-trioxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1,3-diazinan-5-ide
SMILES [NH4+].O=C1NC(=O)[C-](N=C2C(=O)NC(=O)NC2=O)C(=O)N1

Specifications

Melting Point 300°C
Color Red
Physical Form Powder/Solid
Quantity 5 g
Formula Weight 284.19g/mol
Packaging Glass Jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.