missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sodium Phosphate Dibasic Tetrahydrate, ≥99.0% (NT), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 1 kg |
| Linear Formula | NaNH4HPO4 · 4H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 209.07g/mol |
| Percent Purity | ≥99.0% (NT) |
| Packaging | Poly bottle |
| Density | 1.54g/cm3 |
| Chemical Name or Material | Ammonium sodium phosphate dibasic tetrahydrate |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion