missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Succinate, MP Biomedicals

Product Code. 11411446 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411446 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11411446 Supplier MP Biomedicals Supplier No. 0520504980
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 2226-88-2
Quantity 100 g
Molecular Formula C4H12N2O4
Synonym ammonium succinate, diammonium succinate, unii-dtz9pec9jl, ammonium succinate 1:x, butanedioic acid, diammonium salt, dtz9pec9jl, succinic acid, ammonium salt, succinic acid diammonium salt, caswell no. 046, butanedioic acid, ammonium salt 1:2
InChI Key NHJPVZLSLOHJDM-UHFFFAOYSA-N
SMILES C(CC(=O)[O-])C(=O)[O-].[NH4+].[NH4+]
IUPAC Name diazanium;butanedioate
Molecular Weight (g/mol) 152.15
PubChem CID 160625
Formula Weight 152.15
Chemical Name or Material Ammonium Succinate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.