missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfamate, ≥98.0%, ACS Reagent, Solstice

Product Code. 15662280 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15662280 1 kg Poly Bottle 1kg
15652280 100 g Poly Bottle 100g
15672280 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15662280 Supplier Honeywell-Fluka Supplier No. 2287451KG
  • ONLINE EXCLUSIVE PRICE
    284.00 €
    256.08 € / 1kg
    Save 27.92 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7773-06-0
Molecular Formula H6N2O3S
Molecular Weight (g/mol) 114.119
MDL Number MFCD00011429
InChI Key GEHMBYLTCISYNY-UHFFFAOYSA-N
Synonym ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
PubChem CID 24482
ChEBI CHEBI:81950
IUPAC Name azanium;sulfamate
SMILES [NH4+].NS(=O)(=O)[O-]

Specifications

Melting Point 131°C to 135°C
Color White
Physical Form Crystalline
Quantity 1 kg
Linear Formula NH4SO3NH2
UN Number UN3077
Formula Weight 114.12g/mol
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Poly Bottle
Residue after Ignition ≤0.10%
Appearance Crystalline
Density 1g/cm3
Chemical Name or Material Ammonium Sulfamate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.