missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfamate, ACS reagent

Product Code. 10055740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10055740 100 g Plastic Bottle 100g
10301501 5 g Glass Bottle 5g
10075690 500 g Plastic Bottle 500g
10195553 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10055740 Supplier Thermo Scientific Acros Supplier No. 423391000
  • ONLINE EXCLUSIVE PRICE
    44.40 €
    41.60 € / 100g
    Save 2.80 €
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7773-06-0 | H6N2O3S | 114.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7773-06-0
Molecular Formula H6N2O3S
Molecular Weight (g/mol) 114.119
MDL Number MFCD00011429
InChI Key GEHMBYLTCISYNY-UHFFFAOYSA-N
Synonym ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
PubChem CID 24482
ChEBI CHEBI:81950
IUPAC Name azanium;sulfamate
SMILES [NH4+].NS(=O)(=O)[O-]

Specifications

Melting Point 132.0°C to 134.0°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Packaging Plastic Bottle
Residue after Ignition 0.10% max.
Linear Formula H2NSO3NH4
Merck Index 15, 551
Formula Weight 114.12
Percent Purity ≥98.0%
Grade ACS Reagent
Chemical Name or Material Ammonium sulfamate, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium sulfamate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.