missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfamate, Puriss. p.a., ≥99.0% (T), Solstice

Product Code. 15650930 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15650930 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15650930 Supplier Honeywell-Fluka Supplier No. 09960100G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a.,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 131°C to 135°C
CAS 7773-06-0
Color White
pH 4.5 to 6.0 (at 114.1g/L 25°C)
Physical Form Crystalline
Quantity 100 g
Molecular Formula H6N2O3S
Linear Formula NH4SO3NH2
MDL Number MFCD00011429
UN Number UN3077
Synonym ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
InChI Key GEHMBYLTCISYNY-UHFFFAOYSA-N
SMILES [NH4+].NS(=O)(=O)[O-]
IUPAC Name azanium;sulfamate
Molecular Weight (g/mol) 114.119
PubChem CID 24482
ChEBI CHEBI:81950
Formula Weight 114.12g/mol
Percent Purity ≥99.0% (T)
Grade puriss. p.a.
Packaging Poly bottle
Residue after Ignition ≤0.02% (as SO4)
Chemical Name or Material Ammonium Sulfamate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonium sulphamidate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium sulphamidate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.