missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate (2:1), TRC

Product Code. 30137881
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137881 25 g 25g
30008366 100 g 100g
30004216 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137881 Supplier Toronto Research Chemicals Supplier No. TRCA63383525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H2 O4 S . 2 H3 N
Molecular Weight (g/mol) 132.14
Synonym Sulfuric acid ammonium salt (1:2), Sulfuric acid diammonium salt (8CI,9CI), Ammonium sulfate (2:1), Actimaster AMS, Ammonium sulfate, Ammonium sulfate ((NH4)2SO4), Ammonium sulphate, Coaltrol LPA 40, Diammonium sulfate, Diammonium sulphate, Dolamin, Nonnen R 999-10, Para-Go, Polyethylene terephthalate, Sulfuric acid, diammonium salt, Tasker Clear
IUPAC Name azane;sulfuric acid
SMILES N.N.OS(=O)(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)
Formula Weight 132.0205
Chemical Name or Material Ammonium sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.