missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, 98+%

Product Code. 11469173
Change view
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1000g
5000g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469173 5000 g 5000g
11459173 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469173 Supplier Thermo Scientific Alfa Aesar Supplier No. A11682.0I
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Ammonium sulfate is widely utilized as a fertilizer for alkaline soils. In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. As a food additive, it serves as an acidity regulator in flours and breads. It is used in the preparation of other ammonium salts, particularly ammonium persulfate. It is applied in several applications as a flame retardant and a wood preservative. It finds use as raw materials for various chemical products such as dyes and pharmaceutical products. A saturated solution of ammonium sulfate in heavy water is employed as an external standard in sulfur-33 NMR spectroscopy.

Solubility
Soluble in water. Insoluble in alcohol and acetone.

Notes
Incompatible with various substances like highly reactive oxidizing agents and alkalis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name diazanium;sulfate
SMILES N.N.OS(O)(=O)=O

Specifications

Melting Point ∼280°C (decomposition)
Quantity 5000 g
Linear Formula (NH4)2SO4
Merck Index 14,555
Solubility Information Soluble in water. Insoluble in alcohol and acetone.
Formula Weight 132.14
Percent Purity ≥98%
Density 1.769
Chemical Name or Material Ammonium sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.