missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, 99.95% (metals basis)

Product Code. 11443760
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
This item is not returnable. View return policy

Product Code. 11443760

Brand: Thermo Scientific Alfa Aesar 089363.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol

Ammonium sulfate is widely utilized as a fertilizer for alkaline soils. In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. As a food additive, it serves as an acidity regulator in flours and breads. It is used in the preparation of other ammonium salts, particularly ammonium persulfate. It is applied in several applications as a flame retardant and a wood preservative. It finds use as raw materials for various chemical products such as dyes and pharmaceutical products. A saturated solution of ammonium sulfate in heavy water is employed as an external standard in sulfur-33 NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Melting Point ∽280°C (decomposition)
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.95% (metals basis)
Solubility Information Soluble in water and insoluble in acetone.
Formula Weight 132.14
Density 1.769 g/mL
Chemical Name or Material Ammonium sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.