missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, 99+%, ACS reagent

Product Code. 10105563 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Unit Size:
1kg
25g
25kg
500g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10105563 5 kg Plastic bottle 5kg
10556792 25 g Plastic bottle 25g
10697561 500 g Plastic bottle 500g
10556602 1 kg Plastic bottle 1kg
15109293 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10105563 Supplier Thermo Scientific Acros Supplier No. 423400050
  • ONLINE EXCLUSIVE PRICE
    178.00 €
    172.00 € / 5kg
    Save 6.00 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
SMILES N.N.OS(O)(=O)=O

Specifications

Melting Point 280.0°C
Color Silver to Gray
pH 5.0 to 6.0 (5% soln. at 25°C)
Physical Form Granules, Fillings or Chips
Quantity 5 kg
Assay Percent Range 99+%
Linear Formula (NH4)2SO4
Merck Index 15, 552
Solubility Information Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
IUPAC Name diazanium;sulfate
Formula Weight 132.14
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic bottle
Residue after Ignition 0.005 % max.
Density 1.76g/mL
Chemical Name or Material Ammonium sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.