missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, Puratronic™, 99.999% (metals basis)

Product Code. 11390829
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11390829 10 g 10g
11300839 50 g 50g
11310839 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11390829 Supplier Thermo Scientific Alfa Aesar Supplier No. 010639.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol

In biochemistry it is used for purification proteins via ammonium sulfate precipitation method. Ammonium sulfate is used in agriculture as fertilizer; it is a slow releasing nitrogenous fertilizer and a rich source of sulphur nutrient. It is employed as a dough conditioner in bread products. It finds use as raw materials for various chemical products such as dyes and pharmacetical products. It is a component of flame-proofing agents and fire extinguisher powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
SMILES N.N.OS(O)(=O)=O

Especificaciones

Melting Point ∼280°C (decomposition)
Color White
Physical Form Crystalline
Quantity 10 g
Linear Formula (NH4)2SO4
Merck Index 14,555
Solubility Information Highly soluble in water. It is insoluble in acetone,ether and ethanol. It is also insoluble in liquid ammonia.
IUPAC Name sulfuric acid diamine
Formula Weight 132.14
Percent Purity 99.999%
Grade Puratronic
Density 1.769 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium sulfate
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.