missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, TraceSELECT™, Honeywell™

Product Code. 15670930 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 15670930

Brand: Honeywell-Fluka 09979100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TraceSELECT™, for trace analysis,≥99.9999% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7783-20-2
Quantity 100 g
Molecular Formula H8N2O4S
Linear Formula (NH4)2SO4
MDL Number MFCD00003391
UN Number NONH for all modes of transport
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
Solubility Information Completely Soluble in water , 132g/L at 20°C
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILES N.N.OS(O)(=O)=O
IUPAC Name diazanium;sulfate
Molecular Weight (g/mol) 132.13
PubChem CID 6097028
ChEBI CHEBI:62946
Formula Weight 132.14g/mol
Percent Purity ≥99.99% (metals basis)
Grade TraceSELECT
Density 1.77g/mL at 25°C
Chemical Name or Material Ammonium Sulfate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.