Learn More
Ammonium sulfate, ultrapure, 99+%
Descrizione
Ammonium sulfate is a widely used reagent in molecular biology and chromatography. It is useful in the isolation and purification of enzymes and proteins. It is used for the precipitation or fractionation of proteins or for purification of antibodies. It is also useful for crystallographic analysis of nucleic acids and proteins. Ammonium sulfate is also widely used in HPLC of proteins, such as in hydrophobic interaction chromatography. Ammonium sulfate is used in long PCR buffer, in PCR lysis solution, and in secondstrand cDNA synthesis. Also it is used for the HPLC analysis of phospholipids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ammonium sulfate is a widely used reagent in molecular biology and chromatography. It is useful in the isolation and purification of enzymes and proteins. It is used for the precipitation or fractionation of proteins or for purification of antibodies. It is also useful for crystallographic analysis of nucleic acids and proteins. Ammonium sulfate is also widely used in HPLC of proteins, such as in hydrophobic interaction chromatography. Ammonium sulfate is used in long PCR buffer, in PCR lysis solution, and in secondstrand cDNA synthesis. Also it is used for the HPLC analysis of phospholipids.
Solubility
Soluble freely in water, Insoluble in alcohol and acetone.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifica
Specifica
| Melting Point | ∽280°C (decomposition) |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 100 g |
| Density | 1.769 |
| Linear Formula | (NH4)2SO4 |
| Merck Index | 14,555 |
| Solubility Information | Soluble freely in water,Insoluble in alcohol and acetone. |
| Formula Weight | 132.14 |
| Percent Purity | ≥99% |
| Vedi altri risultati |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.