missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15538214 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
Unit Size:
5kg
Each
This item is not returnable. View return policy

Product Code. 15538214

Brand: Avantor J.T.Baker 402702

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Protein Precipitation, Liquid Chromatography and Molecular Biology Applications

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name diazanium;sulfate
SMILES N.N.OS(O)(=O)=O

Specifications

Quantity 1 kg
Formula Weight 132.14
Grade Ultrapure Bioreagent
Packaging Poly Bottle
Specific Gravity 1L = 1.77kg
Chemical Name or Material Ammonium Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.