missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrachloropalladate(II), 98%

Product Code. 10470471 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 10470471

Brand: Thermo Scientific Acros 195170020

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13820-40-1 | Cl4H8N2Pd | 284.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13820-40-1
Molecular Formula Cl4H8N2Pd
Molecular Weight (g/mol) 284.30
MDL Number MFCD00010884
InChI Key IRKVTUVFHGUMMN-UHFFFAOYSA-L
Synonym ammonium tetrachloropalladate ii, ammonium palladium chloride, diammonium tetrachloropalladate 2-, ammonium chloropalladite, bisammonium tetrachloropalladate 2-, diammonium tetrachloro palladite, ammonium tetrachloropalladate, ccris 7828, palladate 2-, tetrachloro-, diammonium, sp-4-1, ammoniumpalladiumchloride
PubChem CID 61682
ChEBI CHEBI:60149
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pd++]

Specifications

Color Black
Physical Form Powder
Quantity 2 g
Assay Percent Range 98%
Linear Formula (NH4)2PdCl4
Fieser 03,81; 01,778; 02,303; 04,379; 05,496; 06,442; 09,344; 10,297; 11,389; 12,367; 13,103; 15,245; 16,259; 17,255
Solubility Information Solubility in water: soluble
IUPAC Name diazanium;tetrachloropalladium(2-)
Formula Weight 284.28
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 2.17
Density 2.1700g/mL
Chemical Name or Material Ammonium tetrachloropalladate(II)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonium tetrachloropalladate(II)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.