Learn More
Ammonium tetrathiotungstate
Description
Ammonium tetrathiotungstate is used to classic building block for transition metal sulfide cluster compounds was recently used in a low-temperature, solid state synthesis of a new series of heterometallic sulfide clusters stabilized with dppm [bis(diphenylphosphino)methane] ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Color | Yellow |
| Physical Form | Chunks and/or Powder |
| Quantity | 2 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 348.07 |
| Odor | Like rotten eggs |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Ammonium tetrathiotungstate |
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.