missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiocyanate, 98+%

Product Code. 11463354
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463354 5000 g 5000g
11453354 250 g 250g
11443354 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463354 Supplier Thermo Scientific Alfa Aesar Supplier No. A10632.0I
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1762-95-4 | CH4N2S | 76.117 g/mol

Ammonium thiocyanate has many uses in manufacturing chemicals such as metal thiocyanates [CuSCN, Ca(SCN)2, NaSCN, KSCN] and in a number of pharmaceuticals. It is used in freezing solutions, pickling, printing, and as a corrosion inhibitor against acid gases. It is used in the photography industry as a stabilizer or an accelerator. It is also used in fertilizers, fabric dyeing, zinc coating, electroplating and as a separator of zirconium and hafnium as well as gold and iron. It is also used in analytical chemistry for titrimetric analysis and to determine the iron content by colorimetry in soft drinks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium thiocyanate has many uses in manufacturing chemicals such as metal thiocyanates [CuSCN, Ca(SCN)2, NaSCN, KSCN] and in a number of pharmaceuticals. It is used in freezing solutions, pickling, printing, and as a corrosion inhibitor against acid gases. It is used in the photography industry as a stabilizer or an accelerator. It is also used in fertilizers, fabric dyeing, zinc coating, electroplating and as a separator of zirconium and hafnium as well as gold and iron. It is also used in analytical chemistry for titrimetric analysis and to determine the iron content by colorimetry in soft drinks.

Solubility
Soluble in water, ethanol, methanol, ammonia and acetone. Insoluble in chloroform and ethyl acetate.

Notes
Light sensitive and hygroscopic. Incompatible with strong oxidizing agents, strong acids and lead nitrate.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Melting Point 147°C to 149°C
Density 1.305
Odor Odorless
Linear Formula NH4SCN
Quantity 5000 g
Beilstein 3595135
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,561
Solubility Information Soluble in water,ethanol,methanol,ammonia and acetone. Insoluble in chloroform and ethyl acetate.
Formula Weight 76.12
Percent Purity ≥98%
Chemical Name or Material Ammonium thiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.