missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, 99.99% Trace Metals Basis, Honeywell™

Product Code. 15655510 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15655510 50 g Poly Bottle 50g
15645510 250 g Poly Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15655510 Supplier Honeywell Research Chemicals Supplier No. 43135450G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

99.99% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Melting Point 140°C to 149°C
Color White
pH 4.5 to 6
Physical Form Solid
Quantity 50 g
Linear Formula NH4SCN
Beilstein 3595135
Solubility Information In water 1630g/L at 20°C
Formula Weight 76.12g/mol
Percent Purity 99.99% (trace metals basis)
Packaging Poly Bottle
Appearance Crystals
Density 1.3g/cm3
Chemical Name or Material Ammonium thiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.