missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiocyanate, 99+%, extra pure

Product Code. 10553761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
10kg
1kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10553761 1 kg Plastic bottle 1kg
10144930 500 g Plastic bottle 500g
10414415 2.5 kg Plastic bottle 2.5kg
15189233 10 kg Plastic Drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10553761 Supplier Thermo Scientific Acros Supplier No. 206500010
  • ONLINE EXCLUSIVE PRICE
    90.55 €
    84.90 € / 1kg
    Save 5.65 €
    6% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1762-95-4 | CH4N2S | 76.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Melting Point 149.0°C
Color White
pH 4.0 to 6.0 (5% aq. soln.)
Physical Form Crystals
Quantity 1 kg
Packaging Plastic bottle
Specific Gravity 1.3
Assay Percent Range 99% min. (on dried basis) (Argentometry)
Density 1.3000g/mL
Linear Formula NH4SCN
Merck Index 15,558
Solubility Information Solubility in water: 1600g/L (20°C). Other solubilities: soluble in alcohol, acetone and ammonia, practically insoluble in chcl3 and ethyl acetate
Formula Weight 76.12
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Ammonium thiocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonium thiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.