missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiocyanate, ACS, 97.5% min

Product Code. p-3256893
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11353308

Brand: Thermo Scientific Alfa Aesar 036247.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fertilizers, photography, fabric dyeing, zinc coating, electroplating, and as a separator of Zr and Hf | CAS: 1762-95-4 | CH4N2S | 76.117 g/mol

Ammonium thiocyanate has many uses in manufacturing chemicals such as metal thiocyanates [CuSCN, Ca(SCN)2, NaSCN, KSCN], and in a number of pharmaceuticals. It is used in freezing solutions, pickling, printing, and corrosion inhibitor against acid gases. It is used in photography industry as a stabilizer or accelerator. Used in fertilizers, fabric dyeing, zinc coating, electroplating, and as a separator of Zr and Hf, also Au and Fe.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Melting Point 147°C to 149°C
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 97.5% min
Beilstein 3595135
Solubility Information Soluble in water,ethanol,methanol,ammonia,and acetone. Practically insoluble in CHCl3and ethyl acetate.
Formula Weight 76.12
Grade ACS
Sensitivity Hygroscopic; Light sensitive
Density 1.305
Chemical Name or Material Ammonium thiocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonium thiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium thiocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.