missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, Solstice

Product Code. 15683240
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
6 x 500 g
40 kg
6 x 1 kg
Unit Size:
100g
1kg
250g
40kg
500g
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15683240 250 g 250g
15663240 100 g 100g
15603250 500 g 500g
15673240 1 kg 1kg
15623250 6 x 500 g Pack of 6
15613250 6 x 1 kg Pack of 6
15693240 40 kg 40kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15683240 Supplier Honeywell-Fluka Supplier No. 31120250G
  • ONLINE EXCLUSIVE PRICE
    54.00 €
    48.47 € / 250g
    Save 5.53 €
    10% Off
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99% (AT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Quantity 250 g
Linear Formula NH4SCN
Beilstein 3595135
Formula Weight 76.12g/mol
Percent Purity ≥99% (AT)
Grade ACS/puriss. p.a.
Residue after Ignition ≤0.02% (as SO4)
Chemical Name or Material Ammonium Thiocyanate, Reag. Ph. Eur. reag. ISO
compliance-icons
Product Identifier
  • Ammonium thiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.