missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Trifluoroacetate, Honeywell Fluka™

Product Code. 15621580 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15621580 50 g 50g
15611580 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15621580 Supplier Honeywell-Fluka Supplier No. 1783950G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for HPLC,≥99.0% (NT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3336-58-1
Molecular Formula C2H5F3NO2
Molecular Weight (g/mol) 132.06
MDL Number MFCD00012615,MFCD03095537
InChI Key YCNIBOIOWCTRCL-UHFFFAOYSA-O
Synonym ammonium trifluoroacetate, azanium 2,2,2-trifluoroacetate, ammoniumtrifluoroacetate, ammonium triflouroacetate, cf3coonh4, acmc-1cq68, 2,2,2-trifluoroacetic acid, azamethane, acetic acid, trifluoro-, ammonium salt 8ci 9ci
PubChem CID 86601334
IUPAC Name azanium;2,2,2-trifluoroacetic acid
SMILES [NH4+].OC(=O)C(F)(F)F

Specifications

Quantity 50 g
Linear Formula CF3CO2NH4
UN Number NONH for all modes of transport
Solubility Information Soluble in water 0.1g/mL at 25°C
Formula Weight 131.05g/mol
Percent Purity ≥99% (NT)
Chemical Name or Material Ammonium Trifluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.