missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Vanadium Oxide, TRC

Product Code. 30012126
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012126 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012126 Supplier Toronto Research Chemicals Supplier No. TRCA6340955G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7803-55-6
Quantity 5 g
Molecular Formula O3 V . H4 N
Synonym Ammonium Vanadate(V) ((NH4)VO3), Vanadic Acid (HVO3), Ammonium Salt, Ammonium Metavanadate, Ammonium Metavanadate (NH4VO3), Ammonium Monovanadate, Ammonium Trioxovanadate, Ammonium trioxovanadate(1-) Ammonium Vanadate, Ammonium Vanadate ((NH4)VO3), Ammonium Vanadate (VO31-), Ammonium Vanadium Oxide ((NH4)VO3) Ammonium Vanadium Trioxide
InChI Formula InChI=1S/H3N.3O.V/h1H3;;;;/q;;;-1;/p+1
SMILES [NH4+].[O-][V](=O)=O
IUPAC Name azanium;oxido(dioxo)vanadium
Molecular Weight (g/mol) 116.98
Formula Weight 116.9631
Chemical Name or Material Ammonium Vanadium Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.