missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin, TRC

Product Code. 17941205
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17941205 1 g 1g
17934415 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17941205 Supplier Toronto Research Chemicals Supplier No. TRCA6345281G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26787-78-0
Molecular Formula C16 H19 N3 O5 S
Molecular Weight (g/mol) 365.4
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-, (2S,5R,6R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[2-amino-2-(p-hydroxyphenyl)acetamido]-3,3-dimethyl-7-oxo-, D- (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(2R)-amino(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-, (2S,5R,6R)- (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[amino(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-, [2S-[2α,5α,6β(S*)]]-, (2S,5R,6R)-6-[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[D(-)-α-Amino-p-hydroxyphenylacetamido]penicillanic acid, 6-[D-(-)-2-Amino-2-(p-hydroxyphenyl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[D-(-)-p-Hydroxy-α-aminobenzyl]penicillin, 6-[D-α-Amino-α-(4-hydroxyphenyl)acetamido]penicillanic acid, A-Gram, AM 73, Abdimox, Acimox, Actimoxi, Adbiotin, Agerpen, Alfamox, Almodan, Alphamox, Amagesen Solutab, Amcill, Amo-flamsian, Amocilline, Amoclen, Amodex, Amoflux, Amolin, Amopen, Amopenixin, Amoram, Amosine, Amox, Amoxa, Amoxal, Amoxapen, Amoxaren, Amoxcillin, Amoxi, Amoxi-Mast, Amoxi-basan, Amoxibiotic, Amoxicilina, Amoxicillin, Amoxicillin Chewable Tablets, Amoxidal, Amoxiden, Amoxidin, Amoxihexal, Amoxil, Amoxillin, Amoxin, Amoxipen, Amoxipenil, Amoxisol, Amoxivan, Amoxivet, Amoxtrex, Amoxy, Amoxy-diolan, Amoxycillin, Amoxypen, Ampidroxyl, Ampy-Penyl, Anemol, Anemolin, Apitart, Apo-Amoxi, Ardine, Ardine (pharmaceutical), Aspenil, Audumic, Azillin, BLP 1410, BRL 2333, Bactamox, Bactox, Betamox, Bimox, Bintamox, Biomox, Bioxidona, Bioxyllin, Blumox, Bridopen, Bristamox, Cabermox, Cemoxin, Cilamox, Clamox, Clamoxyl, Coamoxin, Comoxyl, D-(-)-α-Amino-p-hydroxybenzyl penicillin, D-(α-Amino-p-hydroxybenzyl)penicillin, D-2-Amino-2-(4-hydroxyphenyl)acetamidopenicillanic acid, D-Amoxicillin, Damoxicil, Delacillin, Draximox, Efpenix, Efpinex, Eupen, Eupensol, Excillin, Farconcil, Fisamox, Flemoxin, Foxolin, Fullcilina, Gemox, Gimalxina, Gomcillin, Gramidil, Grinsil, Grinsul, Grunamox, Helvamox, Hiconcil, Hidramox, Hipen, Histocillin, Hosboral, Ibiamox, Ikamoxil, Imacillin, Imaxilin, Imox, Imoxil, Intermox, Isimoxin, Izoltil, Jerramcil, Kamoxin, Lamoxy, Larocilin, Larotid, Limox, Majorpen, Matasedrin, Maxcil, Medimox, Meixil, Metafarma capsules, Metifarma capsules, Mopen, Morgenxil, Mox, Moxacin, Moxaline, Moxarin, Moxcil, Moxilen, Moxlin, Moxylin, Moxypen, Moxyvit, NSC 277174, Novabritine, Novamox, Novamoxin, Novenzymin, Optium, Ospamox, Pamocil, Pamoxicillin, Pasetocin, Penamox, Penbiosyn, Penimox, Pensyn, Piramox, Polymox, Posmox, Protexillin, Rancil, Ranmoxy, Ranoxyl, Reloxyl, Rhamoxilina, Robamox, Rocillin, Ronemox, Saltermox, Samosillin, Samthongcillin, Sawacillin, Sawamezin, Senox, Sia-mox, Sigmopen, Sil-A-mox, Simoxil, Simplamox, Specillin, Sumox, Superpeni, Suprapen, Suramox, Suramox 50, Suramox PM, Teramoxyl, Tolodina, Triafamox, Triamoxil, Trilaxin, Trimox, Twicyl, Unicillin, Utimox, Velamox, Vetomoxin, Vetramox, Virgoxillin, Widecillin, Winmox, Wymox, Yisulon, Zamocillin, Zamox, Zamoxil, Zerrsox, Zimox, p-Hydroxyampicillin
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccc(O)cc3)C(=O)N2[C@H]1C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H19N3O5S/c1-16(2)11(15(23)24)19-13(22)10(14(19)25-16)18-12(21)9(17)7-3-5-8(20)6-4-7/h3-6,9-11,14,20H,17H2,1-2H3,(H,18,21)(H,23,24)/t9-,10-,11+,14-/m1/s1
Formula Weight 365.1045
Chemical Name or Material Amoxicillin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.