missing translation for 'onlineSavingsMsg'
Learn More

Thermo Fisher Scientific Amphipol A8-35

Product Code. 17449292
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
17449292 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 17449292

Brand: Thermo Fisher Scientific A50943

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An amphipathic polymer ideally suited to improve solubilization and stabilization of membrane proteins while maintaining structural integrity and activity.

  • An amphipathic polymer ideally suited to improve solubilization and stabilization of membrane proteins while maintaining structural integrity and activity
  • Detergent-free substitute to extract and stabilize membrane proteins in aqueous solutions
  • Surfactant with dense distribution of hydrophobic chains that have a high affinity for transmembrane surfaces
  • Enables protein solubilization with minimal disruption of membrane environment
  • Facilitates membrane protein structure/folding
  • Suitable for protein solubilization and stabilization for single particle cryo-EM analysis
TRUSTED_SUSTAINABILITY

Specifications

Color White
Molecular Formula (C6.2H10.3O1.35N0.65Na0.35)∼72
Quantity 100 mg
Solubility Information Up to 20% in water
Formula Weight 9 kDa
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.