missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Amphotericin B

Product Code. 18769596
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18769596 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18769596 Supplier R&D Systems™ Supplier No. 6930/100
  • ONLINE EXCLUSIVE PRICE
    317.17 €
    285.60 € / 100mg
    Save 31.57 €
    9% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Forms monovalent ion channels in lipid bilyers

Amphotericin B forms nonselective monovalent ion channels within lipid bilayers. Amphotericin B binds ergosterol in the cell membrane. Antifungal and antiprotozoal.

For more information about how Amphotericin B may be used, see our protocol:

3D Culture of Lung Alveolar Cells
TRUSTED_SUSTAINABILITY

Specifications

CAS 1397-89-3
Quantity 100 mg
Solubility Soluble to 20 mM in DMSO
Product Line Tocris
Molecular Formula C47H73NO17
Formula Weight 924.09
Product Type Antifungal
Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.