missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin sodium salt, Thermo Scientific™

Ampicillin sodium salt, CAS # 69-52-3, is the sodium salt form of ampicillin, a semisynthetic derivative of aminopenicillin compound that shows antibacterial activity against both gram-positive and gram-negative bacteria.

79.00 € - 118.00 €

Chemical Identifiers

CAS 69-52-3
Molecular Formula C16H21N3NaO4S
Molecular Weight (g/mol) 374.411
MDL Number MFCD00064313
InChI Key BSFVNXCYXDYHOD-ZQDFAFASSA-N
PubChem CID 131673879
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15465979
View Documents
Thermo Scientific Alfa Aesar
J63807.06
79.00 €
5g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15475979
View Documents
Thermo Scientific Alfa Aesar
J63807.09
10g
118.00 €
10g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• Ampicillin sodium salt is a semi-synthetic derivative of penicillin with a broad-spectrum antibacterial activity against several species, including Bacillus subtilis, Staphylococcus aureus, Escherichia coli, and Ralstonia solanacearum

• It can inhibit bacterial cell wall synthesis by binding to penicillin-binding proteins, inhibiting peptidoglycan synthesis, which is an essential component of the bacterial cell wall

Application

• Ampicillin sodium salt has been used as a reagent for transformed cells that express β-lactamase

• It can be used to select for ampicillin resistance in mutated and transformed cells

• Ampicillin sodium salt acts as a structural analog of acyl-D-alanyl-D-alanine, acylates the transpeptidase enzyme preventing the cross-linking of the cell wall peptidoglycan, which is necessary for the growth of the bacterium

Specifications

Chemical Identifiers

69-52-3
374.411
BSFVNXCYXDYHOD-ZQDFAFASSA-N
(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
C16H21N3NaO4S
MFCD00064313
131673879
[HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na]

Specifications

69-52-3
C16H21N3NaO4S
4119211
Freely soluble in water; Sparingly soluble in acetone
[HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na]
374.411
371.39
White
MFCD00064313
14,586
BSFVNXCYXDYHOD-ZQDFAFASSA-N
(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
131673879
Ampicillin sodium salt
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P260-P271-P272-P280g-P285-P302+P352-P304+P340-P314-P333+P313-P342+P311-P363-P501c

H317-H334

missing translation for 'einecsNumber' : 200-708-1

missing translation for 'rtecsNumber' : XH8400000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.