missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ampicillin trihydrate

Commonly used to select for ampicillin resistance in mutated and transformed cells

Brand:  Thermo Scientific™ J66514.14

Quantity 25g

Product Code. 15417909

  • 69.40 € / 25g
 Restricted Item
In Stock
Edge
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

7177-48-2
White
>110°C (230°F)
MFCD00072036
684°C
RXDALBZNGVATNY-CWLIKTDRSA-N
(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate
23565
403.45
Nearly Odorless
25g
C16H25N3O7S
5399534
Soluble in water. Practically insoluble in ethanol
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.O.O.O
403.45
CHEBI:31209
Ampicillin trihydrate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.