missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human AMPK alpha-2/beta-1/gamma-3, GST Tag, His Tag Recombinant Protein
Human AMPK alpha-2/beta-1/gamma-3, GST Tag, His Tag Recombinant Protein
520.00 € - 7980.00 €
Specifications
For Use With (Application) | Kinase Assay |
---|---|
Molecular Weight (g/mol) | 89.4/57.5/81.4 kDa |
KinaseFamily | CAMKL Kinase Family |
Protein Length | Full Length |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17138725
|
Invitrogen™
A32857 |
100 μg |
2435.00 €
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930273
|
Invitrogen™
A30487 |
10 μg |
520.00 €
10µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17158765
|
Invitrogen™
A33347 |
1 mg |
7980.00 €
1mg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
AMPK (AMP-activated protein kinase) serine/threonine kinase is a heterotrimeric complex, which acts as sensor of cellular energy levels. The signaling cascades initiated by activating AMPK are critical to regulation of metabolic events in the liver, skeletal muscle, heart, adipose tissue, and pancreas. This AMPK consists of the alpha2, beta1 and gamma3 subunits. While the purified AMPK (A2/B1/G3) complex can be used in a number of different applications, it has been specifically validated for use with our Z’-LYTE™ assay technology to identify novel kinase inhibitors.- Validated for use with the Z’-LYTE™ assay
- Available in multiple pack sizes for convenient use
Specifications
Kinase Assay | |
CAMKL Kinase Family | |
Human | |
GST Tag, His Tag | |
Dry Ice | |
Liquid |
89.4/57.5/81.4 kDa | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Full Length, Recombinant, Active |