missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amrinone, 98%

Product Code. 15494979
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494979 1 g 1g
15404989 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494979 Supplier Thermo Scientific Chemicals Supplier No. J62655.03
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phosphodiesterase III inhibitor and inhibits platelet aggregation | CAS: 60719-84-8 | C10H9N3O

Phosphodiesterase III inhibitor and inhibits platelet aggregation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60719-84-8
Molecular Formula C10H9N3O
Synonym 5-Amino-(3,4′-bipyridin)-6(1H)-one

Specifications

Quantity 1 g
Description Yellow
Content And Storage 2°C to 8°C
For Use With (Application) Phosphodiesterase III inhibitor and inhibits platelet aggregation
Formula Weight 187.20g/mol
Melting Point 294°C to 297°C (dec.)
Solubility Insoluble in water or chloroform; Slightly soluble in acetic acid or DMF; Soluble in DMSO
Product Type Signal Transduction Reagent
Form Crystals
Assay 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.