missing translation for 'onlineSavingsMsg'
Learn More

Amtolmetin, TRC

Product Code. 30339543
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339543 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339543 Supplier Toronto Research Chemicals Supplier No. TRCA63535510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amtolmetin
CAS 87344-05-6
Molecular Formula C17H18N2O4
Quantity 10 mg
Synonym N-[2-[1-Methyl-5-(4-methylbenzoyl)-1H-pyrrol-2-yl]acetyl]glycine, N-[[1-Methyl-5-(4-methylbenzoyl)-1H-pyrrol-2-yl]acetyl]glycine, MED 5, McN 4366, Tolmetin Glycine Amide
InChI Formula InChI=1S/C17H18N2O4/c1-11-3-5-12(6-4-11)17(23)14-8-7-13(19(14)2)9-15(20)18-10-16(21)22/h3-8H,9-10H2,1-2H3,(H,18,20)(H,21,22)
SMILES Cc1ccc(cc1)C(=O)c2ccc(CC(=O)NCC(=O)O)n2C
IUPAC Name 2-[[2-[1-methyl-5-(4-methylbenzoyl)pyrrol-2-yl]acetyl]amino]acetic acid
Molecular Weight (g/mol) 314.34
Formula Weight 314.1267
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.